Skim Milk Agar Plates
Supplier: CULTURE MEDIA &SUPPLIES INC SE
CM575-P25
470175-918PK 32.95 USD
470175-918
Skim Milk Agar Plates
Media for Microbiology Dehydrated Media for Microbiology
For differentiation of organisms based on proteolysis of casein and coagulation.