Stage Micrometers
Supplier: Avantor
470175-914EA 23.45 USD
470175-914
Stage Micrometers
Micrometers
Helpful microscope tool.
  • Ruled 2 x 0.01 mm (wm) slide
  • Shows 2 mm scale in 0.01 mm subdivisions
  • Mounted on a regular glass slide
Use this stage micrometer to calibrate your microscope eyepiece reticles and objective powers.