Carnivorous Plant Set
Supplier: Boreal Laboratories
869100
470175-816KT 64.99 USD
470175-816
Carnivorous Plant Set
Specimens Live Specimens Plants - Living Material
Observe Different Plants’ Methods of Capture and Feeding.
  • Observe carnivorous plants
  • Compare capture and feeding techniques
Three types of carnivorous plants are included in this set, allowing students to compare the various ways they capture insects for food. Plants are also available separately.
This set of plants includes: 1 Purple Pitcher plants (Sarracenia purpurea), 2 Venus Fly Traps (Dionaea sp.) and 1 Sundew (Drosera sp.), 1 live piece of sphagnum moss, and soil for potting in your own containers.
Ordering information: These products contain living or perishable materials. Please specify a delivery date at order time.