Ward's® Bioassay — Effects of Environmental Toxins Lab Activity
Supplier: Avantor
470175-680EA 126.99 USD
470175-680
Ward's® Bioassay — Effects of Environmental Toxins Lab Activity
Educational Classroom Kits and Activities
Conduct Reference Toxicity Tests
  • Grow and analyze buttercrunch lettuce
  • Design a bioassay experiment
  • Includes a teacher's guide
Sites of spills and contamination are constantly identified. Help students understand how toxicity tests are performed in areas of environmental distress with this practical lab. Students test the toxicity of salt on the growth of Buttercrunch lettuce seeds, which are easy to culture and require no maintenance between experiments. Applying their results to environmental problems, students then design another bioassay experiment. Includes a teacher’s guide, student copymasters, and materials for five setups.