Miniature Bulb
Supplier: Norman Lamps
131
470175-414PK 11.15 USD
470175-414
Miniature Bulb
Lights Incandescent Lights, Lamps and Bulbs
Voltage: 1.30V Current: 0.10A Light Output: 0.03 MSCP Type: G3 1/2 Miniature Bulb Base Type: E10 Miniature Screw Base Life Hours: 50 Dimensions: 0.94" L x 0.463" D.