Polypropylene Buchner Funnel
Supplier: United Scientific Supplies
57105-PK/12
470174-670CS 250.85 USD
470174-670
Polypropylene Buchner Funnel
Funnels Filtration Funnels Büchner Funnels
Useable With or Without Filter Paper.
  • Uses 7.0 cm diameter filter paper
Two-piece HDPE funnel with sealed-in, polyethylene filter plate of coarse porosity offers good retention and fast flow under vacuum.