Straight Connecting Tubes
Supplier: Dynalon
227235
470174-240CS 394.95 USD
470174-240
Straight Connecting Tubes
Fittings and Connectors
These autoclavable straight connectors are molded of polypropylene and have serrations to hold tubing tightly and prevent slipping. They have excellent chemical and temperature resistance.