Ten-Well Polyethylene Test Tube Rack
Supplier: Dynalon
259845
470174-092CS 151.5 USD
470174-092
Ten-Well Polyethylene Test Tube Rack
Racks Tube Racks
The chemical resistant, lightweight polyethylene racks can withstand temperatures up to 110°C. The wells are rounded and hold tubes up to 18 mm in dia. Size: 9 3/4"L x 2 3/8"W x 3"H.