Microcentrifuge Tubes
Supplier: Dynalon
399325
470173-924CS 81.3 USD
470173-924
Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
These microcentrifuge tubes are made of polypropylene. The attached lid provides a tight, leakproof seal.
  • Leak tested to 20000 RCF for 20 minutes at 20 °C with an aqueous solution
  • Lid membrane is uniformly thick to ensure trouble free piercing
  • With frosted writing area and graduations