Tray/Tote Box
Supplier: Dynalon
419244
470173-908CS 305.75 USD
470173-908
Tray/Tote Box
Boxes ESD-Safe Boxes
Designed to nest together without sticking, this tray is molded of hefty linear polyethylene.
  • It will not corrode or mildew
  • Size: 21L×12W×6"D
Remains chemically resistant to alkalis, oils, acids, and detergents.