Lead in Soil Test Kit
Supplier: INDUSTRIAL TEST SYSTEMS INC SE
480311
470173-836CS 493.95 USD
470173-836
Lead in Soil Test Kit
Standards Environmental Analysis Standards
Rare Test for Economical Price
  • Test for a common contaminant
  • Quick and simple to use
  • Instructions included
Simply make a slurry with your soil sample using the included reagents, then dip the slurry, and read the results. The detection level is 0–400 ppm. The kit includes instructions and enough materials for five tests.