Sterile Specimen Container
Supplier: MCKESSON GENERAL MEDICAL CORP
491102
470173-818CS 414.2 USD
470173-818
Sterile Specimen Container
Sample Containers General Purpose Containers
Translucent specimen containers with imprinted, screw-on, leak-resistant lid.
  • Tamper-evident seal
  • Includes patient label
  • Not intended for use in pneumatic tubes
  • Sterile inner surface only
  • Single patient use
  • Not made with natural rubber latex
Two graduation scales are marked in 10 cc (mL) and 1/4 oz. increments up to 120 cc. The leak-proof lid has room to write subject information on. Capacity: 4 oz.