VWR® Septa Sealing Mat
Supplier: VWR International
ES53939VWR
470166-590CS 1174.82 USD
470166-590
VWR® Septa Sealing Mat
Seals Microplate Seals
This 96-well septa sealing mat is a cost-effective alternative and is compatible with the ABI 3100.
  • 96-well sealing mat with septa
  • Round well
  • Non-perforated
The septa sealing mat is compatible for use on the ABI 3100. Standard packaging includes 10 mats/pack, 5 packs/case. In addition to the standard packaging option, it is also available in packs of 10.