Chemical Analysis Using Titrations
Supplier: Ward's Science
IS2515
470165-510EA 93.2 USD
470165-510
Chemical Analysis Using Titrations
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
With this kit, students learn how acid-based titrations operate.
  • Activities Identify Concentration of Unknown Acids in Solutions
  • Students Learn How Acid-Based Titrations Operate
  • Required Items Included
  • Kit Uses Developers I, II, III, and IV
A major challenge for any scientist is identifying what and how much of something is in a solution. One of the most common measurement techniques used is titration.
The activities in this kit focus on how acid-base titrations work and how to identify the concentration of an unknown acid. It includes required items and uses Developers I, II, III, and IV.
Ordering information: This kit includes sodium hydroxide, hydrochloric acid, phenolphthalein indicator, a universal indicator, universal indicator color charts, and reaction vessels.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.