Three-Piece Graduated Glass Cylinder Set
Supplier: Wards
CH0346SET
470165-436EA 35.8 USD
470165-436
Three-Piece Graduated Glass Cylinder Set
Cylinders Measuring Cylinders
Made of Heavy-Duty Borosilicate Glass.
  • 10 mL capacity, 0.2 mL graduations
  • 50 mL capacity,1 mL graduations
  • 100 mL capacity, 1 mL graduations
All cylinders include a plastic guard and a detachable plastic hexagonal base.