Premium Square Bosshead
Supplier: Wards
PRBH05
470165-430EA 9.95 USD
470165-430
Premium Square Bosshead
Clamps Bossheads
All Metal Construction to Carry Heavy Loads and Prevent Torque Damage
  • Threaded metal one-piece clamping screws
  • Able to hold flat and round objects up to 15 mm thick
  • Holds rods orthogonal (90°) to one another