Premium Diecast Bosshead
Supplier: Wards
PRBH01
470165-422EA 14 USD
470165-422
Premium Diecast Bosshead
Clamps Bossheads
Threaded Metal One-Piece Clamping Screws
  • Able to hold rods up to 15 mm in diameter
  • Holds rods orthogonal (90°) to one another
  • All metal construction to carry heavy loads and prevent torque damage