Premium Three-Finger Long Clamp with Bosshead, Vinyl Coated
Supplier: Wards
PRCMP07BH
470165-412EA 31.8 USD
470165-412
Premium Three-Finger Long Clamp with Bosshead, Vinyl Coated
Clamps General Purpose Clamps
Vinyl Lined Extension Lab Clamp
  • Maximum clamp opening is 8.5 cm.
  • Bosshead accommodates up to 19 mm diameter rod
Clamp constructed of powder coated steel, three rubber coated fingers attached to a high strength alloy swivel bosshead for ease of use in the lab.