Danger Flammable Sign, EMEDCO
Supplier: EMEDCO
80903GGHECP
470165-306EA 35.4 USD
470165-306
Danger Flammable Sign, EMEDCO
Signs
This environmentally friendly danger safety sign is 100% recyclable as a number two plastic.
  • Ideal for LEED Accreditation
  • Perfect for Indoor or Outdoor Use
  • High-Density Post-Consumer Recycled (PCR) Polyethylene Construction
  • Chemically Resistant To Solvents, Acids, and Alkalis
  • With UV-Inhibitors
Made from high-density post-consumer recycled (PCR) polyethylene with UV-inhibitors, this danger sign is an important reminder in any laboratory setting.
Chemically resistant to solvents, acids, and alkalis, this sign has outstanding durability. It’s perfect for indoor or outdoor use for up to three years. The sign has 3/16" mounting holes for easy hanging.