CENCO® Drilled Ball Set
Supplier: VWR International
470164-714
470164-714EA 110.75 USD
470164-714
CENCO® Drilled Ball Set
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
High Quality.
  • Every set comes in a convenient plastic storage case, with a foam bottom that holds each ball firmly in place
Each set contains 2 each of brass, aluminum, steel, wood, cork, and copper balls. A 3mm hole is drilled through the center of each ball. A slightly larger hole is drilled half way through the center of the ball. This allows the knot of a string to be held at the center of mass of the ball for superior peformance in oscillation experiments.