Plastic Beaker Set, PMP
Supplier: United Scientific Supplies
BMSET5
470164-038EA 61.95 USD
470164-038
Plastic Beaker Set, PMP
Beakers
This durable set of beakers includes five different sizes.
  • Graduated Markings
  • Five Different Sizes
  • Polymethylpentene (PMP) Construction
  • Durable
  • Clear Walls
Made of polymethylpentene, the beakers offer glass-like clarity with the durability of plastic. 
All beakers in the set have graduated markings.
Ordering information: This set includes five graduated PMP beakers in sizes 50 mL, 100 mL, 250 mL, 500 mL, and 1000 mL.