Digital Gauss Meter
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SEA20
470164-000EA 515.05 USD
470164-000
Digital Gauss Meter
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A useful tool for measuring the magnetic field strength for experiments. The range of the probe is 200 and 2kG with a resolution of 1G at 0 - 200 G. The included probe is a hall probe, which is a flat style probe to fit into small gaps.