Air Puck Hovercraft
Supplier: American Scientific
3569-00
470163-906EA 26.05 USD
470163-906
Air Puck Hovercraft
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Demonstrate Newton’s Laws and the conservation of energy and momentum with the amazing new Air Puck!
  • Teacher's guide included, with NGSS alignment
  • Requires 4 × AA batteries, not included
The Air Puck hovercraft is perfectly balanced and glides on a cushion of air for nearly frictionless motion across any smooth surface. Sturdy bumpers will survive collisions with walls, shoes, or other Air Pucks. Use it to demonstrate Newton's First Law, uniform linear motion. Or, collide two Air Pucks to demonstrate conservation of momentum in two dimensions!
Ordering information: Mini hovercraft launcher is recommended but not included.