Deluxe Trundle Wheel
Supplier: American Scientific
6578-00
470163-874EA 89 USD
470163-874
Deluxe Trundle Wheel
Measuring and Leveling Tools
A trundle wheel makes long distances easy to measure.
  • Measures up to 99,999 meters
  • Includes storage case
Roll the wheel along the ground to measure linear distance. A metered counter measures up to 99,999 meters, heavy-duty construction, telescoping handle for comfort, built-in stand which folds in, rubber wheel trim for accuracy and convenient carrying storage case.