Acid Rain Science Fair Kit
Supplier: Ward's Science
IS5032
470163-314KT 18.85 USD
470163-314
Acid Rain Science Fair Kit
Educational Materials Biology Educational Materials Intro to Science Learning Activities
Determine if You have Acid Rain in Your Area.
  • Great field study
  • Ideal for younger students
  • All required chemicals are included
The Acid rain test kit is great to use in the classroom or for science fair projects to determine if you have acid rain in your area. Collect water samples and use a low range pH indicator to determine the pH of your water sample with a colorimetric comparison chart.
Ordering information: Includes Low Range pH Indicator, Test Tubes, Color Chart, and Instructions.