Ward's® Chemistry Up In Vapor Demonstration
Supplier: Ward's Science
IS7033
470163-150EA 41.6 USD
470163-150
Ward's® Chemistry Up In Vapor Demonstration
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
"Genie in the tube."
  • Safely show a decomposition reaction in the classroom or lab
  • Demonstration can be performed 5 times with materials provided
Observe the dramatic catalytic decomposition of hydrogen peroxide to produce oxygen, water vapor, and an abundance of heat. This dramatic, large-scale demo illustrates concepts of catalysts, decomposition reactions, and exothermic reactions. Glass tube sold separately.
Ordering information: This kit includes hydrogen peroxide, sodium iodide, and an instruction manual with SDS. A glass tube is needed but not included.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.