TLC Chromatography Paper
Supplier: Ward's Science
AD-CHROM-PK/1
470163-136EA 36.9 USD
470163-136
TLC Chromatography Paper
Filters Filter Paper Chromatography Paper
An economical supply for your chromatography studies.
  • One plastic backed sheet
  • Easy to cut and use for an entire classroom
This thin TLC plate is perfect for use with chromatography experiments. Forty 2×5 cm TLC sheets can be made from this 20×20 cm sheet, enough to support a whole lab of students.