The Molar Volume of a Gas
Supplier: Ward's Science
IS8026
470163-116KT 30.6 USD
470163-116
The Molar Volume of a Gas
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
How Much is One Mole?
  • Classic AP Chemistry Experiment
  • Instructions Included
Students will determine the volume of one mole of gas. This is accomplished by generating a known mass of oxygen gas, measuring its temperature, volume and pressure, and then using the data to calculate the molar volume at STP. The oxygen is generated by the decomposition of potassium chlorate at high temperature.
Includes:
Potassium Chlorate
Manganese Dioxide