Synthesis Of Aspirin
Supplier: Ward's Science
IS8021
470163-108KT 55.15 USD
470163-108
Synthesis Of Aspirin
Educational Materials Chemistry Educational Materials Gas Analysis Learning Activities
Using Organic Chemistry, Can You Make Aspirin?
  • Classic AP Chemistry Experiment
  • Instructions Included
In a reaction vessel salicylic acid, acetic anhydride and sulfuric acid are mixed. The exothermic reaction will cause the temperature to increase to 70-80°C. Once the reaction is complete the vessel is cooled in an ice bath and the acetylsalicylic acid crystallizes out. This material is re-crystallized in toluene to purify the product. The product identity is confirmed by melting point. The mass of the re-crystallized product is used to determine the yield of the synthesis. In this experiment, we will perform the synthesis of acetylsalicylic acid (aspirin), purification by re-crystallization and use melting point to confirm identity.
Ordering information: Kit includes Acetic Anhydride, Salicylic Acid, Sulfuric Acid, and Toluene.