Mole Ratio Of Reactants
Supplier: Ward's Science
IS8003
470163-072KT 46.55 USD
470163-072
Mole Ratio Of Reactants
Educational Materials Chemistry Educational Materials Microchemistry Learning Activities
Stoichiometry for AP Chemistry
  • Classic AP Chemistry Experiment
  • Instructions Included
  • Determine the perfect ratio!
Using the method of continuous variation, two solutions are combined in various ratios. To select the ratio that produces the most product or consumes the most reactants, students must find an empirical method which is proportional to the amount of reaction that occurs. The reaction selected for this experiment is exothermic and the optimum ratio produces the greatest temperature change. In this experiment the total numbers of moles of reactants are kept constant while varying each reactant. The measurements are made on each different ratio until the optimum ratio, the stoichiometric ratio in the equation, is made which consumes the greatest amount of reactants, produces the greatest amount of product and produces the greatest amount of heat.
Includes:
Sodium Hypochlorite
Sodium Thiosulfate, anhydrous