Standard Grade Qualitative Filter Papers
Supplier: Shiv Dial Sud & Sons
FP-001-185 FP-001-215 FP-001-240 FP-002-090 FP-002-110 FP-002-125 FP-002-150 FP-001-090 FP-001-110 FP-001-125 FP-002-185 FP-002-240 FP-003-090 FP-003-110 FP-003-125 FP-003-150 FP-003-215 FP-003-240 FP-001-150
470162-956CS 978.2 USD
470162-956 470162-958 470162-960 470162-962 470162-964 470162-966 470162-968 470162-996 470163-000 470163-002 470163-004 470163-008 470163-010 470163-012 470163-014 470163-016 470163-020 470163-022 470163-024
Standard Grade Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
Qualitative Filter Paper is available in three standard grades.
  • Applicable to Most Science Education Applications
  • Grades One, Two, and Three Available
  • Grade One Works with Routine Applications
  • Grade Two Filters Large Samples
  • Grade Three Works with Buchner Funnels
Grade One filter papers are mainly used for routine applications such as separation of precipitates and filtering liquids. They have medium retention and flow rates. Grade Two filter papers are more retentive and absorbent than Grade One papers. Usually, Grade Two filter papers have an 8 µm particle size range and are used for filtering larger sample types such as soil.
Grade Three filter papers are approximately double the thickness of Grade One filter paper, which makes them more durable and suitable for use with Buchner funnels.
Packaging: One pack has 100 papers, and one case has 4800 papers.