Digital Melting Point Apparatus
Supplier: Dynalon
DMP100
470162-898EA 1536.65 USD
470162-898
Digital Melting Point Apparatus
Melting Point Apparatus
Ideal for Use in Educational Chemistry.
  • Magnifying lens that allows a view of up to two samples illuminated clearly by a built in LED light.
  • Adjustable Slow Speed temperature ramp rate from 1°C to 10°C.
  • Fast Stage temperature set that saves time prior to carrying out the melt procedure.
  • Metal feet that extend and help provide an appropriate viewing angle.
The DMP100 makes measurement simple. It can be an indispensable, space-saving, bench top tool for the modern science education laboratory and for select industrial application areas such as quality control testing for the purity of pharmaceutical and other organic crystalline solids. The LED temperature display and “Read" button that captures the melt temperature makes the DMP100 easy to use. The DMP100 includes an Operations Manual with detailed use instructions and one pack of 100 glass capillary tubes, open on one end.