Power Function Generator
Supplier: American 3B Scientific
1009956
470162-794EA 1476.8 USD
470162-794
Power Function Generator
Educational Materials Math Educational Materials Measurement Learning Activities
Compact function generator
  • With retractable feet.
  • Includes power supply
  • Input voltage 110 V
Featuring illuminated, digital display for frequency, signal form, offset and other parameters. The output is short-circuit protected as well as being protected against induced voltages and spark discharges, for when experiment leads are unintentionally pulled out while coils are connected. In internal sweep mode, one trigger pulse is output per cycle and the voltage output is proportional to the frequency.