DC Power Supply 0 to 500 V
Supplier: American 3B Scientific
1003307
470162-724EA 1162.49 USD
470162-724
DC Power Supply 0 to 500 V
Power Supplies
Low voltage power supply with four outputs primarily intended to simultaneously supply power for the Teltron line of electron tubes and Helmholtz Coils, with four independent, adjustable DC voltages. The DC voltages are stabilized, regulated, floating, and galvanically isolated from one another, in addition to being short circuit proof and secure from external voltages.