Helmholtz Coils Demonstration Series
Supplier: American 3B Scientific
1000644
470162-712EA 723.49 USD
470162-712
Helmholtz Coils Demonstration Series
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Generate a uniform magnetic field perpendicular to the axis of the Teltron D series tubes.
Teltron Tube Holder D allows the coils to be configured in the Helmholtz geometry or to produce magnetic fields aligned vertically, or coaxial, with respect to the tube axis. The maximum field strength generated by the coils (~ 10 mT) is approximately 300 times weaker than that produced by an MRI machine (~ 3T), which are known to be safe for use by people with medical implants such as pacemakers. Required but not included is the tube holder.