Electron Tube Holder Demonstration Series
Supplier: American 3B Scientific
1008507
470162-710EA 536.89 USD
470162-710
Electron Tube Holder Demonstration Series
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Tube holder is made of robust plastic for holding D-series electron tubes. Stand has a wishbone, 360° rotating clamp made of heat-resistant plastic and two base holes for affixing the Teltron Helmholtz Coils. On rubber feet to prevent slipping.
Specifications:
Dimensions: approx. 230x175x320 mm³
Weight: approx. 1.5 kg