Polypropylene Funnels, Dynalon
Supplier: Dynalon
412334-0002
470161-302CS 256.25 USD
470161-302
Polypropylene Funnels, Dynalon
Funnels Filtration Funnels Filter Funnels
These Polypropylene Funnels are chemical and acid resistant and can be sterilized. These funnels are offered in four different diameters/capacities to meet all your funneling needs.