Mohr Buret Pinchcock Attachment
Supplier: Avantor
470161-250EA 27.55 USD
470161-250
Mohr Buret Pinchcock Attachment
Burets
Designed for the Flint Glass Mohr Burets. Consists of a short piece of rubber tubing, glass buret tip, and a pinchcock clamp. Easily slips over the end of any buret not having a stopcock.