Medium Wall Latex Tubing
Supplier: Primeline
034RB-50'
470161-184EA 81.2 USD
470161-184
Medium Wall Latex Tubing
Tubing Transfer Tubing
Connector tubing for many applications.
  • Two lengths available
  • Two colors available
This medium wall latex connector tubing is nonsterile and has only 50 mg or less of water extractable protein per gram. This is important since proteins in natural rubber can cause severe allergic reactions to those individuals who are allergic to these types of protein.