GPI Anatomicals® Osteoarthritis Hand Model
Supplier: GPI ANATOMICALS
1930
470160-246EA 144.85 USD
470160-246
GPI Anatomicals® Osteoarthritis Hand Model
Models Human Biology Models
This is full size right hand with cutaway views to reveal effects of osteoarthritis.
  • Removable from base for examination
  • Keycard mounted on base
Full size right hand with cutaway views to reveal effects of osteoarthritis, including: osteophytes (bone spurs), Heberden’s and Bouchard’s nodes, and swan neck deformity of the thumb. Shows other anatomy affected by osteoarthritis: ligaments, tendons, muscle, and cartilage. For comparison, normal anatomy is also shown.