Transformer Accessory Kit
Supplier: Wards
PH0849AK2
470157-638EA 72.75 USD
470157-638
Transformer Accessory Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate Lenz's Law!
  • Excellent accessory to a variable gap magnet
Lenz’s law transformer accessory kit to demonstrate the production of large magnetic fields and lenz’s law damping on pendulums with and without slits.