Antacid Lab Activity
Supplier: Avantor
470154-080KT 142.39 USD
470154-080
Antacid Lab Activity
Microchemistry Kits
Discover How Antacids Work
Using microchemistry techniques, students determine the neutralization and buffering characteristics of an over-the-counter antacid (included). Since all tests are performed using microscale techniques, only a small amount of test material is required. The kit contains three types of antacids, instructions, and materials for seven setups.
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.