Etherizer
Supplier: Avantor
470154-018EA 29.95 USD
470154-018
Etherizer
Bottles Drosophila Bottles
Specially designed unit to safely and easily etherize Drosophila or other living materials.
  • Etherized Drosophila drop into a collecting bottle
The bottle containing live material is inverted over the funnel. A few drops of ether are placed on a dispersing wick under the rubber plug.
Delivery information: Includes dropper pipet.