Lab Scoops
Supplier: Bel-Art
F36747-0000_SE
470153-820CS 115.3 USD
470153-820
Lab Scoops
Scoops
Molded polypropylene scoops are rigid, strong, and smooth.
Steam autoclavable at 121°C/250°F. Scoops are 22cc with a 1 1/4" handle.
Delivery information: Package of 12.