Disposable Polygon Spinbar Magnetic Stirring Bars
Supplier: Bel-Art
37112-6016
470153-810BG 282.9 USD
470153-810
Disposable Polygon Spinbar Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Ideal for single use applications mixing radioactive reagents or when cross contamination might be a concern
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.