Fast Release Pi-Pumps
Supplier: Bel-Art
F37904-0025 F37904-0010
470153-802CS 614 USD
470153-802 470153-804
Fast Release Pi-Pumps
Pipettes Pipette Controllers
Designed for fast and accurate one-handed operation.
  • Reduce the risk of injury from repetitive pipetting
  • Expel entire contents in one shot or smaller amounts precisely
Each pump is resistant to acids, bases, and most solvents; and it can be easily disassembled for cleaning.