Coleslaw Chemistry Kit
Supplier: Avantor
470152-860KT 98.69 USD
470152-860
Coleslaw Chemistry Kit
Educational Classroom Kits and Activities
Use Cabbage Juice as a pH Indicator
  • Ideal for Grades 8-11
  • Create Your Own pH Reference Chart
  • 5 Activities Included
This unique chemistry lab is based on the properties of red cabbage juice as a pH indicator. Students make indicator solution and pH strips out of red cabbage juice and then use the strips to create their own pH reference chart. Students test the acidity of "rain" in a controlled atmosphere.
Includes:
Forceps
Safety Matches
3" x 5" Index Cards
Bags
Filter Paper Strips
Glass Plates
Graduated Pipettes
Spot Plates
Buffer Capsules - pH 3 - 11
Instructions
Required but Not Provided: Head of Red Cabbage, Distilled Water, Container to boil cabbage, Stove or hot plate, Knife, Cutting Board, Scissors, Beakers, Dropping Bottles (optional), Colored Pencils or Crayons, Permanent Markers, Plastic Strainer, Vials or ice cube trays, Erlenmeyer flasks with stoppers.