Galileo Kit
Supplier: Avantor
470152-832EA 78.89 USD
470152-832
Galileo Kit
Educational Classroom Kits and Activities
Re-create His Famous Experiment
  • Includes an inclined plane, steel ball, funnel assembly, and instructions
  • A graduated cylinder, support stand, and clamp, required, are available separately
Galileo discovered, using an inclined plane and a ball, that objects fall at a constant rate of acceleration; if the angle of the inclined plane is changed, it does not affect that rate. Now you and your students can observe the same effect in your own classroom.