Density Tube
Supplier: Avantor
470152-828EA 36.99 USD
470152-828
Density Tube
Educational Classroom Kits and Activities
Teacher Developed!
  • Grades 7–12
  • Size: 24.8cm H
Physics teacher Victor Noto has developed a great way to demonstrate heat transfer by convection. His density tube uses water at different temperatures to produce bulk liquid flow. By timing how long it takes for a small volume of liquid to move up to the top of the tube, students can determine if the rate of heat transfer depends on the density differences. Includes instructions.