Pump and Filter Set
Supplier: Penn-Plax
SWK1UL
470151-850CS 1225.75 USD
470151-850
Pump and Filter Set
Specimens Live Specimens Aquaria - Living Material
Complete Equipment for 5- to 10-Gallon Aquariums
The set comes with a quiet-running air pump, filter that fits in a bottom corner of the tank, airstone and water outlet tube, air line tubing, bag of activated carbon, dacron filter floss, and a handbook with instructions on how to set up an aquarium.